Learn More
TAME, MedChemExpress
Código de producto. 16471757
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
50 mg
100 mg
Tamaño de la unidad:
100 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16471757 50 mg 50 miligramos
16446136 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16471757 Proveedor Medchem Express N.º de proveedor HY13255
MedChemExpress TAME is an inhibitor of anaphase-promoting complex/cyclosome (APC/C or APC), which binds to APC/C and prevents its activation by Cdc20 and Cdh1, produces mitotic arrest. TAME is not cell permeable.
TAME is an inhibitor of anaphase-promoting complex/cyclosome (APC/C or APC), which binds to APC/C and prevents its activation by Cdc20 and Cdh1, produces mitotic arrest. TAME is not cell permeable.
Especificaciones
Nombre del producto químico o material TAME
CAS 901-47-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C14H22N4O4S
Cantidad 50 mg
Información de solubilidad DMSO : 100 mg/mL (292.05 mM; Need ultrasonic)
SMILES CC1=CC=C(C=C1)S(N[C@H](C(OC)=O)CCCNC(N)=N)(=O)=O
Peso molecular (g/mol) 342.41
Formula Weight (peso de la fórmula) 342.41
Porcentaje de pureza 99.68%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death