Learn More
TAME hydrochloride, MedChemExpress
Código de producto. 16481757
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
50 mg
100 mg
Tamaño de la unidad:
100 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16481757 50 mg 50 miligramos
16456136 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16481757 Proveedor Medchem Express N.º de proveedor HY13255A
MedChemExpress TAME hydrochloride is an inhibitor of anaphase-promoting complex/cyclosome (APC/C or APC), which binds to APC/C and prevents its activation by Cdc20 and Cdh1, produces mitotic arrest. TAME hydrochloride is not cell permeable.
TAME hydrochloride is an inhibitor of anaphase-promoting complex/cyclosome (APC/C or APC), which binds to APC/C and prevents its activation by Cdc20 and Cdh1, produces mitotic arrest. TAME hydrochloride is not cell permeable.
Especificaciones
Nombre del producto químico o material TAME hydrochloride
CAS 1784-03-8
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C14H23ClN4O4S
Cantidad 50 mg
Información de solubilidad DMSO : 155 mg/mL (409.11 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (263.94 mM)
SMILES CC1=CC=C(C=C1)S(N[C@H](C(OC)=O)CCCNC(N)=N)(=O)=O.[H]Cl
Peso molecular (g/mol) 378.87
Formula Weight (peso de la fórmula) 378.87
Porcentaje de pureza 98.43%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death