Learn More
Talsaclidine, MedChemExpress
Código de producto. 16488147
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
Tamaño de la unidad:
1 miligramo
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16488147 5 mg 5 miligramos
16431319 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16488147 Proveedor Medchem Express N.º de proveedor HY128855
MedChemExpress Talsaclidine is a muscarinic agonist with preferential neuron-stimulating properties. Talsaclidine is a full agonist at the M1 subtype, and as a partial agonist at the M2 and M3 subtypes.
Talsaclidine is a muscarinic agonist with preferential neuron-stimulating properties. Talsaclidine is a full agonist at the M1 subtype, and as a partial agonist at the M2 and M3 subtypes.
Especificaciones
Nombre del producto químico o material Talsaclidine
Forma física Oil
CAS 147025-53-4
Color Yellow
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C10H15NO
Para utilizar con (aplicación) Neuroscience-Neuromodulation
SMILES C#CCO[C@@H]1[C@@H]2CCN(CC2)C1
Peso molecular (g/mol) 165.23
Formula Weight (peso de la fórmula) 165.23
Porcentaje de pureza 97.0%