Learn More
TAK1/MAP4K2 inhibitor 1, MedChemExpress
Código de producto. 16474117
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16474117 50 mg 50 miligramos
16420668 5 mg 5 miligramos
16418228 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16474117 Proveedor Medchem Express N.º de proveedor HY77251
MedChemExpress TAK1/MAP4K2 inhibitor 1 is a potent dual TGFβ-activated kinase 1 (TAK1) and mitogen-activated protein kinase kinase kinase kinase 2 (MAP4K2) inhibitor, with IC50s of 41.1 nM and 18.2 nM, respectively.
TAK1/MAP4K2 inhibitor 1 is a potent dual TGFβ-activated kinase 1 (TAK1) and mitogen-activated protein kinase kinase kinase kinase 2 (MAP4K2) inhibitor, with IC50s of 41.1 nM and 18.2 nM, respectively.
Especificaciones
Nombre del producto químico o material TAK1/MAP4K2 inhibitor 1
CAS 1315330-11-0
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C29H31F3N6O2
Cantidad 50 mg
Información de solubilidad DMSO : 50 mg/mL (90.48 mM; Need ultrasonic)
SMILES CC(N1)=CC2=C1N=CN=C2OC3=C(C)C=CC(C(NC4=CC=C(CN5CCN(CC)CC5)C(C(F)(F)F)=C4)=O)=C3
Peso molecular (g/mol) 552.59
Formula Weight (peso de la fórmula) 552.59
Porcentaje de pureza 98.07%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease