Learn More
Tocris Bioscience™ TAK 21d
Descripción
TAK 21d is a potent FAAH inhibitor; (IC50 values are 0.28 and 0.72 nM, at rat and human FAAH respectively). Displays analgesic effects in vivo models of neuropathic and inflammatory pain. Brain penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-[4-(3,4-Difluorophenyl)-2-pyrimidinyl]-N-3-pyridazinyl-1-piperazinecarboxamide |
| CAS | 1143578-94-2 |
| Cantidad | 50 mg |
| Objetivo | Fatty Acid Amide Hydrolase (FAAH) Inhibitors |
| Fórmula molecular | C19H17F2N7O |
| Formula Weight (peso de la fórmula) | 397.38 |
| Pureza | >98% |