Learn More
T863, MedChemExpress
Código de producto. 16488028
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16488028 10 mg 10 miligramos
16470488 5 mg 5 miligramos
16467806 100 mg 100 miligramos
16433957 50 mg 50 miligramos
16403746 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16488028 Proveedor Medchem Express N.º de proveedor HY32219
MedChemExpress T863 is an orally active, selective and potent DGAT1 (Acyl-CoA:diacylglycerol acyltransferase 1) inhibitor that interacts with the acyl-CoA binding site of DGAT1, and inhibits triacylglycerol synthesis in cells.
T863 is an orally active, selective and potent DGAT1 (Acyl-CoA:diacylglycerol acyltransferase 1) inhibitor that interacts with the acyl-CoA binding site of DGAT1, and inhibits triacylglycerol synthesis in cells.
Especificaciones
Nombre del producto químico o material T863
Forma física Powder
CAS 701232-20-4
Color Blanco
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C22H26N4O3
Para utilizar con (aplicación) Metabolism-sugar/lipid metabolism
Información de solubilidad DMSO : ≥ 50 mg/mL (126.75 mM)
SMILES O=C(O)C[C@H]1CC[C@H](C2=CC=C(C3=NC4=C(N)N=CN=C4OC3(C)C)C=C2)CC1
Peso molecular (g/mol) 394.47
Formula Weight (peso de la fórmula) 394.47
Porcentaje de pureza 95.06%