Learn More
T0901317, MedChemExpress
Código de producto. 16495046
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16495046 100 mg 100 miligramos
16444078 10 mg 10 miligramos
16400437 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16495046 Proveedor Medchem Express N.º de proveedor HY10626
MedChemExpress T0901317 is an orally active and highly selective LXR agonist with an EC50 of 20 nM for LXRα. T0901317 activates FXR with an EC50 of 5 μM. T0901317 is RORα and RORγ dual inverse agonist with Ki values of 132 nM and 51 nM, respectively. T0901317 induces apoptosis and inhibits the development of atherosclerosis in low-density lipoprotein (LDL) receptor-deficient mice.
T0901317 is an orally active and highly selective LXR agonist with an EC50 of 20 nM for LXRα. T0901317 activates FXR with an EC50 of 5 μM. T0901317 is RORα and RORγ dual inverse agonist with Ki values of 132 nM and 51 nM, respectively. T0901317 induces apoptosis and inhibits the development of atherosclerosis in low-density lipoprotein (LDL) receptor-deficient mice.
Especificaciones
Nombre del producto químico o material T0901317
CAS 293754-55-9
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C17H12F9NO3S
Cantidad 100 mg
Información de solubilidad DMSO : 100 mg/mL (207.76 mM; Need ultrasonic)
SMILES OC(C(F)(F)F)(C(F)(F)F)C1=CC=C(N(CC(F)(F)F)S(C2=CC=CC=C2)(=O)=O)C=C1
Peso molecular (g/mol) 481.33
Formula Weight (peso de la fórmula) 481.33
Porcentaje de pureza 99.8%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death