Learn More
Tocris Bioscience™ T 3364366
Description
T 3364366 is a potent and reversible fatty acid desaturase 1 (FADS1; Δ5 desaturase) inhibitor (IC50 values are 1.9 nM and 2.1 nM in human and rat hepatocyte cell lines, respectively). Exhibits >300 fold selectivity for FADS1 over FADS2 (Δ6 desaturase). Inhibits arachidonic acid production in rat and human hepatocyte cell lines. Orally bioavailable.
Specifications
Specifications
| Nombre del producto químico o material | N-[2-[[3,4-Dihydro-4-oxo-3-[4-(2,2,2-trifluoroethoxy)phenyl]thieno[3,4-d]pyrimidin-2-yl]thio]ethyl]acetamide |
| CAS | 1356354-09-0 |
| Cantidad | 10 mg |
| Fórmula molecular | C18H16F3N3O3S2 |
| Formula Weight (peso de la fórmula) | 443.46 |
| Pureza | 98% |