Learn More
MedChemExpress SYBR Green qPCR Master Mix (Low ROX)
Especificaciones
Especificaciones
| Contenido y almacenamiento | Stored at -20°C protecting from light, and is stable for up to 18 months. Avoid repetitive freeze-thaw cycles while using. For immediate use, components may be stored at 2°C to 8°C. |
| Tipo de producto | Kits |
| Cantidad | 500 reacciones |
| Condiciones de envío | Room temperature |