Learn More
Tocris Bioscience™ SW 033291
High affinity 15-PGDH inhibitor; promotes hematopoiesis and hepatocyte proliferation
Marca: Tocris Bioscience™ 5759/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SW 033291 is a high affinity 15-hydroxyprostaglandin dehydrogenase (15-PGDH) inhibitor (Ki = 0.1 nM). Increases PGE2 levels in A549 cells in vitro, as well as in bone marrow, colon, lung and liver in mice. Promotes hematopoiesis, recovery from DSS-induced colitis, and hepatocyte proliferation after partial hepatectomy in mice.Especificaciones
| 2-(Butylsulfinyl)-4-phenyl-6-(2-thienyl)-thieno[2,3-b]pyridin-3-amine | |
| 459147-39-8 | |
| 10 mg |