Learn More
Tocris Bioscience™ Surfen
Descripción
Surfen is a heparan sulfate antagonist. Inhibits heparan sulfate-mediated cell attachment in vitro. Maintains hESCs in a pluripotent state by reversible inhibition of glycan-growth factor interactions.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N,N'-Bis(4-amino-2-methyl-6-quinolinyl)urea dihydrochloride |
| CAS | 5424-37-3 |
| Cantidad | 25 mg |
| Objetivo | Compounds for ESC/iPSC Proliferation and Renewal |
| Fórmula molecular | C21H20N6O0.2 HCl |
| Formula Weight (peso de la fórmula) | 445.35 |
| Pureza | 98% |