Learn More
Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Microtiter Plates, Total V-Bottom, Level 2 High-Throughput Applications
Tienda Thermo Scientific ProductosDescripción
Thermo Scientific™ SureSTART™ Level 2 WebSeal™ 96-Well Total V-Bottom Microtiter Plates are designed to ensure high quality data with an uninterrupted workflow for high-throughput GC and HPLC/UHPLC applications. They are ideal for regulated labs that use water and polar solvents.
- Performance Level 2 Plates for High Throughput Applications
- Ideal for non-regulated labs that use water and polar solvents
- Certified plastic well plates, seals, tapes
- GC-MS and LC-MS tested
- Suitable for HPLC/UHPLC applications
- Suitable for QA/QC workflows and sensitive applications with low sample concentrations
- Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
- Specification Certified
- All Thermo Scientific SureSTART WebSeal well plates are dimensionally verified
Especificaciones
Especificaciones
| Código de barras | Includes Barcode |
| Formato | SBS Standard |
| matriz, red, conjunto | 8 x 12 |
| Línea de productos | SureSTART WebSeal Level 2 |
| Color | Clear |
| Tipo de producto | 96-Well Microtiter Plate |
| Material | Polyethylene |
| N.° de pocillos | 96 |
| Esterilidad | Non-sterile |
| Tratamiento de superficie | Non-coated |
| Mostrar más |