Learn More
Sulindac-d6, TRC
Product Code. 30412995
Change view
Click to view available options
Cantidad:
2.5 mg
Unit Size:
2.50 miligramo
Product Code. Cantidad unitSize
30412995 2.5 mg 2.50 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412995 Supplier Toronto Research Chemicals Supplier No. TRCS6992172.5MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Sulindac-d6
CAS 2714166-73-9
Fórmula molecular C20H11D6FO3S
Cantidad 2.5 mg
Sinónimo (1Z)-5-Fluoro-2-methyl-1-[(4-(methylsulfinyl)phenyl]methylene]-1H-indene-3-acetic Acid-d6, cis-5-Fluoro-2-methyl-1-[(p-methylsulfinyl)benzylidenyl]indene-3-acetic Acid-d6, Aflodac-d6, Algocetil-d6, Arthrocine-d6, Artribid-d6, Citireuma-d6, Clinoril-d6, Clisundac-d6, Imbaral-d6, MK 231-d6, Mobilin-d6, Reumofil-d6, Reumyl-d6, Sudac-d6, Sulindac-d6, Sulindac sulfoxide-d6, Sulinol-d6, Sulreuma-d6, cis-Sulindac-d6
Fórmula InChI InChI=1S/C20H17FO3S/c1-12-17(9-13-3-6-15(7-4-13)25(2)24)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9-/i2D3,9D,11D2
SMILES O=S(C([2H])([2H])[2H])C(C=C1)=CC=C1/C([2H])=C2C(C)=C(C([2H])([2H])C(O)=O)C3=CC(F)=CC=C3/2
Nombre IUPAC Ácido (Z)-2-(5-fluoro-2-metil-1-((4-((metil-d3)sulfinil)fenil)metilen-d)-1H-inden-3-il)acético-2,2-d2
Peso molecular (g/mol) 362.45
Formula Weight (peso de la fórmula) 362.13