Descripción
Immunogen sequence: QIAQLQDFVR AKGSQSGRHL QTHSNTIVVS LQSKLASMSN DFKSVLEVRT ENLKQQRSRR EQFSRAPVSA LPLAPNHLGG GAVVLGAESH ASKDVAIDMM DSRTSQQLQL IDEQDSYIQS RADTMQNIES TIVELGSIFQ QLAHMV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Mediates endoplasmic reticulum to Golgi transport. Together with p115/USO1 and GM130/GOLGA2, involved in vesicle tethering and fusion at the cis-Golgi membrane to maintain the stacked and inter-connected structure of the Golgi apparatus.
Especificaciones
Especificaciones
| Antígeno | STX5 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | STX5 |
| N.º de referencia del gen | Q13190 |
| Alias de gen | 0610031F24Rik; D19Ertd627e; SED5; Stx5; Stx5a; syntaxin 5; syntaxin 5A; syntaxin-5 |
| Símbolos de los genes | STX5 |
| Mostrar más |