Learn More
STEMCELL Technologies STAT5 Inhibitor
Descripción
- STAT5 Inhibitor is a cell-permeable, nonpetidic nicotinoyl hydrazone that specifically suppresses STAT5 (IC50 = 47 μM) via binding to its SH2 domain
- It shows reduced potency towards the SH2 domains of STAT1, STAT3, or LCK (Müller et al.)
- Induces partial apoptosis of mature eosinophils at high concentrations (Schwartz et al.)
- Rescues IL-17 production in a proportion of restimulated TH17 clones (Zielinski et al.)
- Reduces growth of induced regulatory T cells (iTregs) in vitro (Betts et al.)
Especificaciones
Especificaciones
| CAS | 285986-31-4 |
| Cantidad | 5 mg |
| Fórmula molecular | C16H11N3O3 |
| Pureza | ≥95% |