Learn More
Tocris Bioscience™ st-Ht31
Descripción
st-Ht31 is a stearated form of the peptide Ht-31. Inhibits the interaction between the RII subunits of cAMP-dependent PKA and A-kinase anchoring protein (AKAP) in cell extracts. Cell permeable.
Especificaciones
Especificaciones
| CAS | 188425-80-1 |
| Contenido y almacenamiento | Store at -20°C |
| Peso molecular | 2798.27 |
| Cantidad | 1 mg |