Learn More
Tocris Bioscience™ SSR 180711 hydrochloride
Descripción
SSR 180711 hydrochloride is a partial agonist at α7 nAChR. Enhances episodic memory and reverses MK-801 -induced deficits in retention of episodic memory. Increases extracellular dopamine levels in rat prefrontal cortex. Also exhibits antidepressant-like properties.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Bromophenyl 1,4-diazabicyclo[3.2.2]nonane-4-carboxylate hydrochloride |
| CAS | 446031-79-4 |
| Cantidad | 10 mg |
| Objetivo | Nicotinic (a7) Receptor Agonists |
| Fórmula molecular | C14H17BrN2O2.HCl |
| Formula Weight (peso de la fórmula) | 361.66 |
| Pureza | >98% |