Learn More
SSEA-5 Mouse, anti-Human, FITC, Clone: 8e11, STEMCELL Technologies
Código de producto. 17188791
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17188791 100 Tests 100 tests
17198791 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17188791 Proveedor Stemcell Technologies N.º de proveedor 60063FI
Mouse Monoclonal Antibody
  • The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
  • Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
  • The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures
Especificaciones
Antígeno SSEA-5
Aplicaciones Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 8e11
Conjugado FITC
Especie del huésped Mouse
Cantidad 100 Tests
Primario o secundario Primary
Especies diana Human
Isotype IgG1 κ
Tipo de célula Pluripotent Stem Cells