missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRSF8 Polyclonal Antibody

Descripción
Antibody detects endogenous levels of total SFRS2B.
The SR (serine/arginine-rich) family contains a number of phosphoproteins that function as essential and alternative splicing factors. The SR family of proteins is characterized by the presence of a ribonucleoprotein (RNP)-type RNA binding motif and a carboxyl-terminal arginine-serine-rich (RS) domain. SFRS2B is a member of the SR family and functions as an essential splicing factor in vitro.The SR (serine/arginine-rich) family contains a number of phosphoproteins that function as essential and alternative splicing factors. The SR family of proteins is characterized by the presence of a ribonucleoprotein (RNP)-type RNA binding motif and a carboxyl-terminal arginine-serine-rich (RS) domain. The protein encoded by this gene is a member of the SR family and functions as an essential splicing factor in vitro. This gene is thought to be an expressed PR264/SC35 retropseudogene.
Especificaciones
Especificaciones
| Antígeno | SRSF8 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | SRSF8 |
| N.º de referencia del gen | Q9BRL6 |
| Alias de gen | DSM-1; pre-mRNA-splicing factor SRP46; serine and arginine rich splicing factor 8; serine/arginine-rich splicing factor 8; SFRS2B; Splicing factor SRp46; Splicing factor, arginine/serine-rich 2B; splicing factor, arginine/serine-rich, 46kD; SR splicing factor 8; SRP46; SRSF8 |
| Símbolos de los genes | SRSF8 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?