missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRRM1 Polyclonal Antibody

Descripción
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 92%; Zebrafish: 91%.
Part of pre- and post-splicing multiprotein mRNP complexes. Involved in numerous pre-mRNA processing events. Promotes constitutive and exonic splicing enhancer (ESE)-dependent splicing activation by bridging together sequence-specific (SR family proteins, SFRS4, SFRS5 and TRA2B/SFRS10) and basal snRNP (SNRP70 and SNRPA1) factors of the spliceosome. Stimulates mRNA 3'-end cleavage independently of the formation of an exon junction complex. Binds both pre-mRNA and spliced mRNA 20-25 nt upstream of exon-exon junctions. Binds RNA and DNA with low sequence specificity and has similar preference for either double- or single-stranded nucleic acid substrates.
Especificaciones
Especificaciones
| Antígeno | SRRM1 |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | SRRM1 |
| N.º de referencia del gen | Q8IYB3 |
| Alias de gen | 160-KD; AA407769; LOW QUALITY PROTEIN: serine/arginine repetitive matrix protein 1; plenty-of-prolines 101; plenty-of-prolines-101; Pop101; Ser/Arg-related nuclear matrix protein; serine and arginine repetitive matrix 1; serine/arginine repetitive matrix 1; serine/arginine repetitive matrix protein 1; Srm160; SR-related nuclear matrix protein of 160 kDa; SRRM1; wu:fb34f04; wu:fb38b06; zgc:63802 |
| Símbolos de los genes | SRRM1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?