Learn More
Tocris Bioscience™ SR 9238
Descripción
SR 9238 is a potent and selective LXR inverse agonist (IC50 values are 43 and 214 nM for LXRβ and LXRα, respectively). Selectively inhibits liver LXR over peripheral LXR. Exhibits selectivity for LXR over a panel of 20 other nuclear receptors, including FXR. Reduces hepatic steatosis in obese mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Ethyl 5-[[[[3′-(Methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl][(2,4,6-trimethylphenyl)sulfonyl]amino]methyl]-2-furancarboxylate |
| CAS | 1416153-62-2 |
| Cantidad | 50 mg |
| Objetivo | LXR-like Receptor Antagonists |
| Fórmula molecular | C31H33NO7S2 |
| Formula Weight (peso de la fórmula) | 595.73 |
| Pureza | >98% |