Learn More
Tocris Bioscience™ SR 49059
Descripción
Potent and selective non-peptide vasopressin V1A receptor antagonist; devoid of agonist activity. Displays high affinity and efficacy at both rat (Ki = 1.6 nM) and human (Ki = 1.1 - 6.3 nM) V1A receptors. Potently antagonizes arginine vasopressin-induced effects in vitro (IC50 = 3.7 nM for inhibition of human platelet aggregation) and is orally active in vivo.
Especificaciones
Especificaciones
| CAS | 150375-75-0 |
| Cantidad | 10 mg |
| Tipo de producto | SR 49059 |
| Pureza | >99% |
| Peso molecular | 620.498 |