Learn More
SR 16832, Tocris Bioscience™
PPAR gamma Receptor Antagonists
Marca: Tocris Bioscience 6383/5
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SR 16832 is a dual site PPARγ inhibitor. Acts at orthosteric and allosteric sites in the ligand binding domain. Inhibits binding of endogenous ligands and transcriptional activity of PPARγ, more effectively than the orthosteric covalent antagonists GW 9662 (Cat. No. 1508) and T 0070907 (Cat. No. 2301).Especificaciones
| 2088135-12-8 | |
| 2-Chloro-N-(6-methoxy-4-quinolinyl)-5-nitrobenzamide | |
| 5 mg |