Learn More
Tocris Bioscience™ SR 11302
Descripción
Inhibitor of activator protein-1 (AP-1) transcription factor activity that displays antitumor effects in vivo. Does not activate transcription from the retinoic acid response element (RARE) and displays no activity at retinoic acid receptors (EC50 > 1 μM for RARα, RARβ, RARγ and RXRα).
Especificaciones
Especificaciones
| Nombre del producto químico o material | SR 11302 |
| CAS | 160162-42-5 |
| Cantidad | 10 mg |
| Fórmula molecular | C26H32O2 |
| Formula Weight (peso de la fórmula) | 376.54 |
| Pureza | >98% |
| Peso molecular | 376.54 |