Learn More
SQ109, MedChemExpress
Product Code. 16496278
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16496278 10 mg 10 miligramos
16466616 100 mg 100 miligramos
16458867 5 mg 5 miligramos
16442407 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496278 Supplier Medchem Express Supplier No. HY14989
This item is not returnable. View return policy
MedChemExpress SQ109 is a potent inhibitor of the trypomastigote form of the parasite, with IC50 for cell killing of 50±8 nM. SQ109, targets MmpL3, is an antitubercular agent.
SQ109 is a potent inhibitor of the trypomastigote form of the parasite, with IC50 for cell killing of 50±8 nM.
Specifications
Nombre del producto químico o material SQ109
Forma física Liquid
CAS 502487-67-4
Color Colorless
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C22H38N2
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo NSC 722041
Información de solubilidad DMSO : ≥ 25 mg/mL (75.63 mM)
SMILES C/C(C)=C/CC/C(C)=C/CNCCNC1[C@H]2C[C@H]3C[C@@H]1C[C@H](C3)C2
Peso molecular (g/mol) 330.55
Formula Weight (peso de la fórmula) 330.55
Porcentaje de pureza 95.0%