Learn More
SPT Imidazopyridine 1, Tocris Bioscience™
Potent serine palmitoyl transferase inhibitor
Marca: Tocris Bioscience 6055/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SPT Imidazopyridine 1 is a potent serine palmitoyl transferase (SPT) inhibitor (IC50 = 5 nM). Dose-dependently inhibits incorporation of 14C-serine into ceramide. Reduces plasma ceramide levels in vivo. Also increases plasma HDL and reduces vLDL cholesterol levels in rats.Especificaciones
| 4-[6-(1-Oxoheptyl)imidazo[1,2-a]pyridin-8-yl]benzeneacetic acid | |
| 1933533-18-6 | |
| >98% | |
| 50 mg |