Learn More
SPHINX31, MedChemExpress
Código de producto. 16481137
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16481137 50 mg 50 miligramos
16477637 5 mg 5 miligramos
16455666 100 mg 100 miligramos
16444888 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16481137 Proveedor Medchem Express N.º de proveedor HY117661
MedChemExpress SPHINX31 is a potent and selective inhibitor of serine/arginine-rich protein kinase 1 (SRPK1), with an IC50 of 5.9 nM. SPHINX31 inhibits phosphorylation of serine/arginine-rich splicing factor 1 (SRSF1). SPHINX31 is a potential topical therapeutic for neovascular eye disease.
SPHINX31 is a potent and selective inhibitor of serine/arginine-rich protein kinase 1 (SRPK1), with an IC50 of 5.9 nM. SPHINX31 inhibits phosphorylation of serine/arginine-rich splicing factor 1 (SRSF1). SPHINX31 is a potential topical therapeutic for neovascular eye disease.
Especificaciones
Nombre del producto químico o material SPHINX31
Forma física Solid
CAS 1818389-84-2
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C27H24F3N5O2
Para utilizar con (aplicación) Cancer-Kinase/protease
Información de solubilidad DMSO : 17.33 mg/mL (34.15 mM; Need ultrasonic)
SMILES O=C(NC1=C(N2CCN(CC3=CC=CC=N3)CC2)C=CC(C(F)(F)F)=C1)C4=CC=C(O4)C5=CC=NC=C5
Peso molecular (g/mol) 507.51
Formula Weight (peso de la fórmula) 507.51
Porcentaje de pureza 99.12%