Learn More
Tocris Bioscience™ Spexin
Description
Spexin is a potent galanin receptor 2/3 (GAL2/GAL3) agonist (EC50 values are 45.7 and 112.2 nM, respectively). Exhibits no significant activity at galanin receptor 1. Endogenous satiety-inducing peptide; inhibits long chain fatty acid uptake by adipocytes and decreases food consumption in diet-induced obese mice and rats. Attenuates LH secretion in goldfish. Exhibits anxiolytic effects in vivo.
Specifications
Specifications
| CAS | 1370290-58-6 |
| Contenido y almacenamiento | Store at −20°C |
| Para utilizar con (aplicación) | Potent galanin receptor 2/3 (GAL2/GAL3) agonist |
| Liofilizado | Yes |
| Peso molecular | 1619.9 |
| Tipo de producto | Peptide |
| Cantidad | 1 mg |
| Secuencia | NWTPQAMLYLKGAQ |
Para uso exclusivo en investigación.