Learn More
Tocris Bioscience™ SP 10
Descripción
SP 10 is a peptide originally derived from SARS-CoV Spike (S) protein; corresponds to amino acid residues 668 to 679. Highly potent inhibitor of SARS-CoV S protein and ACE2 interaction (IC50 = 1.88 nM in biochemical assay). Inhibits interaction of SARS-CoV S protein and Vero E6 cells in vitro. Please note, this peptide displays 80% sequence identity to homologous site on SARS-CoV-2 S-protein (residues 689-697).
Especificaciones
Especificaciones
| Gene ID (Entrez) | 59272 |
| Pureza | 95% |
| CAS | 882157-88-2 |
| Contenido y almacenamiento | Store at -20°C |
| Alias de gen | ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 |
| Peso molecular | M.W. (Observed): 1315.5 |
| Cantidad | 1 mg |