Learn More
Dietilditiocarbamato-d10 de sodio, TRC
Product Code. 30415590
Change view
Click to view available options
Cantidad:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad
30415590 25 mg
30412783 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30415590 Supplier Toronto Research Chemicals Supplier No. TRCS62420225MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1261395-23-6
Fórmula molecular C5D10NNaS2
Peso molecular (g/mol) 181.32
Sinónimo N,N-Diethyl-carbamodithioic Acid-d10 Sodium Salt, Diethyldithio-carbamic Acid-d10 Sodium Salt, Dithiocarb-d10, Dithiocarb-d10 sodium, Ditiocarb-d10 sodium, Imuthiol-d10, Kupral-d10, Soxinol ESL-d10, Thiocarb-d10
SMILES S=C(S[Na])N(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H]
Specifications
Nombre del producto químico o material Sodium Diethyldithiocarbamate-d10
Cantidad 25 mg
Fórmula InChI InChI=1S/C5H11NS2.Na/c1-3-6(4-2)5(7)8;/h3-4H2,1-2H3,(H,7,8);/q;+1/p-1/i1D3,2D3,3D2,4D2;
Formula Weight (peso de la fórmula) 181.078