Learn More
Butirato de sodio-3,3,4,4,4-d5, CDN
Product Code. 30319171
Change view
Click to view available options
Cantidad:
0.25 g
0.5 g
Unit Size:
0.25 g
0.50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30319171 0.5 g 0.50 g
30321350 0.25 g 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319171 Supplier CDN Supplier No. CDND53710.5G
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Descubra más de 4000 compuestos deuterados de alta calidad, con envío directo desde stock para mayor rapidez. Ofrecemos muchas API únicas, incluidos ingredientes farmacéuticos activos, nitrosoaminas, aminoácidos, esteroides, gases, pesticidas y ácidos grasos.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Sodium Butanoate-3,3,4,4,4-D5
CAS 204244-79-1
Fórmula molecular C4 D5 H2 O2 . Na
Cantidad 0.5 g
Sinónimo Butanoic-3,3,4,4,4-d5 acid, sodium salt (9 CI), Butanoic-d5 acid sodium salt, Sodium butanoate-3,3,4,4,4-d5, Sodium butyrate-d5, Sodium butanoate-3,3,4,4,4-D5
Fórmula InChI InChI=1 S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1/i1D3,2D2;
SMILES [Na+].[2 H]C([2 H])([2 H])C([2 H])([2 H])CC(=O)[O-]
Nombre IUPAC sodio;3,3,4,4,4-pentadeuteriobutanoato
Peso molecular (g/mol) 115.118
Formula Weight (peso de la fórmula) 115.066 g/mol
Porcentaje de pureza 98 atom % D, min 98% Chemical Purity