Learn More
Óxido de aluminio sódico, grado técnico, Thermo Scientific Chemicals
Código de producto. 11323637
Cambiar vista
Click to view available options
Cantidad:
100 g
500 g
Tamaño de la unidad:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
11323637 500 g 500 g
11313637 100 g 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 11323637 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 035453.36
Solicitar formato a granel o personalizado
CAS: 1302-42-7 | Al2O3·Na2O
Sodium aluminum oxide is used in the isomerization of alkene and isomerization of amines. It is also used in water treatment as an adjunct and as a coagulant aid to flocculation. It is involved in the removal of silica and phosphates from waste water. It is employed to accelerate concrete solidification. It finds application in the paper industry and fire brick production. It acts as an intermediate in the production of zeolites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 1302-42-7
Fórmula molecular Al2O3·Na2O
Número MDL MFCD00801010
Sinónimo Sodium aluminate
Especificaciones
Nombre del producto químico o material Sodium aluminum oxide
Color Blanco
Forma física cristalino/a/s
Cantidad 500 g
Información de solubilidad Insoluble in alcohol,soluble in waterInsoluble in alcohol. Soluble in water.
Formula Weight (peso de la fórmula) 163.94
Sensibilidad Hygroscopic
Aspecto White crystalline
Densidad 1.52