missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: LRVRARVADR LYGVYKVHGN YGRVFSEWSA IEKEMGDGLQ SAGHHMDVYA SSIDDILEDE EHYADQLKEY LFYAEALRAV CRKHELMQYD LEMAAQDLAS KKQQCEELVT GTVRTFSLKG MTTKLFGQET PEQREARIKV LEEQINEGE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain. These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases. These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes. The related protein, SNX3, comprises a distinct subgroup of nexins that share less sequence similarity outside of the PX domain and have dramatically different binding affinities for the tyrosine kinase receptors.
Especificaciones
Especificaciones
| Antígeno | SNX4 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | SNX4 |
| N.º de referencia del gen | O95219, Q91YJ2 |
| Alias de gen | 1810036H14Rik; AI037066; ATG24B; Snx4; sorting nexin 4; sorting nexin-4 |
| Símbolos de los genes | SNX4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
