Learn More
Kit TerpiMix 2500 de Smart SolutionsTM μg/mL en isopropanol, Dr. Ehrenstorfer
Especificaciones
Especificaciones
| Nombre del producto químico o material | (-)-alpha-Cedrene∣(-)-Borneol∣(-)-Fenchone∣(-)-Isopulegol∣(-)-Trans-Caryophyllene∣(-)-Verbenone∣(+)-Borneol∣(+)-Carvone∣(+)-Dihydrocarveol∣(+)-Fenchone∣(+)-Fenchyl Alcohol∣(±)-beta-Citronellol∣(1S)-(+)-3-Carene∣(cis + trans)-Nerolidol∣1-Methyl-2-isopropylbenzene∣1-Terpinen-4-ol∣3-Isopropyltoluene (m-Cymol)∣4-Cymene∣4-Thujanol∣Acetic Acid-Octyl Ester∣alpha-(-)-Thujone∣alpha-Bisabolol∣alpha-Farnesene∣alpha-Humulene∣alpha-Phellandrene∣alpha-Terpinene∣alpha-Terpineol∣beta-Ocimene∣beta-Pinene∣Camphene∣Carvacrol∣Caryophyllene Oxide∣Cedrol∣Citral∣Delta-3-Carene∣delta-Valerolactam∣DL-alpha-Pinene∣DL-Camphor∣DL-Linalool∣Eucalyptol (1,8-Cineole)∣Farnesol (Mixture of Isomers)∣gamma-Terpinene∣Geraniol∣Geranyl Acetate∣Guaiol∣Isoborneol∣Isobornyl Acetate∣Limonene∣Menthol Racemic∣Menthone∣Myrcene∣Nerol∣Nootkatone∣Phytane∣Phytol (cis/trans mixture)∣Piperitone∣R-(+)-Pulegone∣Sabinene∣Safranal∣Squalene∣Terpinolene∣Thymol∣Valencene |
| CAS | 469-61-4, 464-45-9, 7787-20-4, 89-79-2, 87-44-5, 1196-01-6 |
| Nombres de analitos o componentes | (-)-alpha-Cedrene∣(-)-Borneol∣(-)-Fenchone∣(-)-Isopulegol∣(-)-Trans-Caryophyllene∣(-)-Verbenone∣(+)-Borneol∣(+)-Carvone∣(+)-Dihydrocarveol∣(+)-Fenchone∣(+)-Fenchyl Alcohol∣(±)-beta-Citronellol∣(1S)-(+)-3-Carene∣(cis + trans)-Nerolidol∣1-Methyl-2-isopropylbenzene∣1-Terpinen-4-ol∣3-Isopropyltoluene (m-Cymol)∣4-Cymene∣4-Thujanol∣Acetic Acid-Octyl Ester∣alpha-(-)-Thujone∣alpha-Bisabolol∣alpha-Farnesene∣alpha-Humulene∣alpha-Phellandrene∣alpha-Terpinene∣alpha-Terpineol∣beta-Ocimene∣beta-Pinene∣Camphene∣Carvacrol∣Caryophyllene Oxide∣Cedrol∣Citral∣Delta-3-Carene∣delta-Valerolactam∣DL-alpha-Pinene∣DL-Camphor∣DL-Linalool∣Eucalyptol (1,8-Cineole)∣Farnesol (Mixture of Isomers)∣gamma-Terpinene∣Geraniol∣Geranyl Acetate∣Guaiol∣Isoborneol∣Isobornyl Acetate∣Limonene∣Menthol Racemic∣Menthone∣Myrcene∣Nerol∣Nootkatone∣Phytane∣Phytol (cis/trans mixture)∣Piperitone∣R-(+)-Pulegone∣Sabinene∣Safranal∣Squalene∣Terpinolene∣Thymol∣Valencene |
| Condiciones de envío | Room Temperature |
| Certificaciones/conformidad | ISO 17034 |
| Solvent or Matrix | Isopropanol |
| Cantidad | 3 x 1 mL |
| Concentration or Composition (by Analyte or Components) | (-)-alpha-Cedrene 2500 μg/mL∣(-)-Borneol 2500 μg/mL∣(-)-Fenchone 2500 μg/mL∣(-)-Isopulegol 2500 μg/mL∣(-)-Trans-Caryophyllene 2500 μg/mL∣(-)-Verbenone 2500 μg/mL∣(+)-Borneol 2500 μg/mL∣(+)-Carvone 2500 μg/mL∣(+)-Dihydrocarveol 2500 μg/mL∣(+)-Fenchone 2500 μg/mL∣(+)-Fenchyl Alcohol 2500 μg/mL∣(±)-beta-Citronellol 2500 μg/mL∣(1S)-(+)-3-Carene 2500 μg/mL∣(cis + trans)-Nerolidol 2500 μg/mL∣1-Methyl-2-isopropylbenzene 2500 μg/mL∣1-Terpinen-4-ol 2500 μg/mL∣3-Isopropyltoluene (m-Cymol) 2500 μg/mL∣4-Cymene 2500 μg/mL∣4-Thujanol 2500 μg/mL∣Acetic Acid-Octyl Ester 2500 μg/mL∣alpha-(-)-Thujone 2500 μg/mL∣alpha-Bisabolol 2500 μg/mL∣alpha-Farnesene 2500 μg/mL∣alpha-Humulene 2500 μg/mL∣alpha-Phellandrene 2500 μg/mL∣alpha-Terpinene 2500 μg/mL∣alpha-Terpineol 2500 μg/mL∣beta-Ocimene 2500 μg/mL∣beta-Pinene 2500 μg/mL∣Camphene 2500 μg/mL∣Carvacrol 2500 μg/mL∣Caryophyllene Oxide 2500 μg/mL∣Cedrol 2500 μg/mL∣Citral 2500 μg/mL∣Delta-3-Carene 2500 μg/mL∣delta-Valerolactam 2500 μg/mL∣DL-alpha-Pinene 2500 μg/mL∣DL-Camphor 2500 μg/mL∣DL-Linalool 2500 μg/mL∣Eucalyptol (1,8-Cineole) 2500 μg/mL∣Farnesol (Mixture of Isomers) 2500 μg/mL∣gamma-Terpinene 2500 μg/mL∣Geraniol 2500 μg/mL∣Geranyl Acetate 2500 μg/mL∣Guaiol 2500 μg/mL∣Isoborneol 2500 μg/mL∣Isobornyl Acetate 2500 μg/mL∣Limonene 2500 μg/mL∣Menthol Racemic 2500 μg/mL∣Menthone 2500 μg/mL∣Myrcene 2500 μg/mL∣Nerol 2500 μg/mL∣Nootkatone 2500 μg/mL∣Phytane 2500 μg/mL∣Phytol (cis/trans mixture) 2500 μg/mL∣Piperitone 2500 μg/mL∣R-(+)-Pulegone 2500 μg/mL∣Sabinene 2500 μg/mL∣Safranal 2500 μg/mL∣Squalene 2500 μg/mL∣Terpinolene 2500 μg/mL∣Thymol 2500 μg/mL∣Valencene 2500 μg/mL |