Learn More
SM-164, MedChemExpress
Código de producto. 16489497
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16489497 5 mg 5 miligramos
16446958 10 mg 10 miligramos
16427036 100 mg 100 miligramos
16413017 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16489497 Proveedor Medchem Express N.º de proveedor HY15989
MedChemExpress SM-164 is a cell-permeable Smac mimetic compound. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains with an IC50 value of 1.39 nM and functions as an extremely potent antagonist of XIAP.
SM-164 is a cell-permeable Smac mimetic compound. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains with an IC50 value of 1.39 nM and functions as an extremely potent antagonist of XIAP.
Especificaciones
Nombre del producto químico o material SM-164
CAS 957135-43-2
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C62H84N14O6
Cantidad 5 mg
Información de solubilidad DMSO : 25 mg/mL (22.29 mM; Need ultrasonic)
SMILES C[C@H](NC)C(N[C@H]1CCCC[C@](CC[C@H]2C(N[C@@H](C3=CC=CC=C3)C4=CN(CCCCC5=CC=C(CCCCN6N=NC([C@@H](NC([C@@H]7CC[C@@](CCCC[C@@H]8NC([C@@H](NC)C)=O)([H])N7C8=O)=O)C9=CC=CC=C9)=C6)C=C5)N=N4)=O)([H])N2C1=O)=O
Peso molecular (g/mol) 1121.42
Formula Weight (peso de la fórmula) 1121.42
Porcentaje de pureza 98.42%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death