Learn More
Tocris Bioscience™ SK 216
Descripción
SK 216 is a plasminogen activator inhibitor-1 (PAI-1) inhibitor. Inhibits VEGF-induced migration and tube formation in HUVECs. Also attenuates TGF-β dependent epithelial-mesenchymal transition and fibroblast to myofibroblast differentiation in vitro. Reduces size and weight of subcutaneous tumors, and inhibits metastasis and angiogenesis in vivo. Reduces bleomycin-induced pulmonary fibrosis in vivo. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-[5-[[6-[5-(1,1-Dimethylethyl)-2-benzoxazolyl]-2-naphthalenyl]oxy]pentyl]propanedioic acid disodium salt |
| CAS | 654080-03-2 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 10 mg |
| Fórmula molecular | C29H29NO6Na2 |
| Formula Weight (peso de la fórmula) | Observed MW: 533.52 |