Learn More
STEMCELL Technologies SIS3
Descripción
- SIS3 (specific inhibitor of SMAD3) is a cell-permeable selective inhibitor of transforming growth factor β1 (TGF-β1)-dependent SMAD3 phosphorylation (IC50 = 3 μM) and SMAD3-mediated cellular signaling (Jinnin et al.) SIS3 has no effect on SMAD2, p38 mitogen-activated protein kinase (MAPK), extracellular signal-regulated kinase (ERK), or phosphoinositide 3-kinases
- Facilitates the production of anterior ectoderm from mouse induced pluripotent stem cells (Oshima et al.)
- Inhibits myofibroblast differentiation of normal human dermal fibroblasts (Jinnin et al.)
Especificaciones
Especificaciones
| CAS | 1009104-85-1 |
| Sinónimo | Smad3 Inhibitor |
| Cantidad | 500 μg |
| Fórmula molecular | C28H27N3O3 |
| Pureza | ≥98% |