Learn More
Shogaol, MedChemExpress
Código de producto. 16423436
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16423436 25 mg 25 miligramos
16416118 10 mg 10 miligramos
16408717 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16423436 Proveedor Medchem Express N.º de proveedor HY14616
MedChemExpress Shogaol ([6]-Shogaol), an active compound isolated from Ginger (Zingiber officinale Rosc), exhibits a variety of biological activities including anticancer, anti-inflammation, and anti-oxidation.
Shogaol ([6]-Shogaol), an active compound isolated from Ginger (Zingiber officinale Rosc), exhibits a variety of biological activities including anticancer, anti-inflammation, and anti-oxidation.
Especificaciones
Nombre del producto químico o material Shogaol
CAS 555-66-8
Color Incolora
Notas de grado de pureza Research
Fórmula molecular C17H24O3
Cantidad 25 mg
Sinónimo [6]-Shogaol 6-Shogaol
Información de solubilidad DMSO : ≥ 50 mg/mL (180.92 mM)
SMILES OC1=C(OC)C=C(CCC(/C=C/CCCCC)=O)C=C1
Peso molecular (g/mol) 276.37
Formula Weight (peso de la fórmula) 276.37
Porcentaje de pureza 98.92%
Grado Research
Forma física Liquid
Para utilizar con (aplicación) Cancer-Kinase/protease