Learn More
Tocris Bioscience™ SHIN 1
Descripción
SHIN 1 is a potent serine hydroxymethyltransferase (SHMT) inhibitor (IC50 values are 5 and 13 nM at SHMT1 and SHMT2, respectively). Inhibits growth of HCT-116 cells in vitro (IC50 = 870 nM). Unstable in liver microsome assays.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 6-Amino-1,4-dihydro-4-[5-(hydroxymethyl)[1,1-biphenyl]-3-yl]-3-methyl-4-(1-methylethyl)pyrano[2,3-c]pyrazole-5-carbonitrile |
| CAS | 2146095-85-2 |
| Cantidad | 5 mg |
| Objetivo | Other Transferase Inhibitors |
| Fórmula molecular | C24H24N4O2 |
| Formula Weight (peso de la fórmula) | 3081 |
| Pureza | 98% |