Learn More
Tocris Bioscience™ SGC SMARCA-BRDVIII
Descripción
SGC-SMARCA-BRDVIII is a selective and high affinity SMARCA2/4 and PB1(bromo 5)-selective SWI/SNF bromodomains (Kd(ITC) values are 35, 36 and 13 nM, respectively). SGC-SMARCA-BRDVIII exhibits selectivity over 25 other bromodomain families and shows no off-target activity on 85 protein kinases screened using temperature-shift binding assays. SGC-SMARCA-BRDVIII suppresses formation of 3T3-L1 mouse fibroblasts into adipocytes (EC50 <1.0 μM).
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1,1-Dimethylethyl 4-[3-amino-6-(2-hydroxyphenyl)-4-pyridazinyl]-1-piperazinecarboxylate bromodomains |
| CAS | 1997319-84-2 |
| Cantidad | 50 mg |
| Objetivo | Bromodomain Inhibitors |
| Fórmula molecular | C19H25N5O3 |
| Pureza | 0.98 |