missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SFTPB Monoclonal Antibody (SPB02 (1B9))
GREENER_CHOICE

Código de producto. 15688018
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15688018 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15688018 Proveedor Invitrogen™ N.º de proveedor MA1204

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Detects mature surfactant protein B (8kDa monomer and 16kDa Dimeric forms) and pro surfactant protein B (24kDa) in western blot. Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 92%; Rabbit: 85%; Rat: 100%; Sheep: 79%.

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventilation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SFTPB
Aplicaciones Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon SPB02 (1B9)
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
génica SFTPB
N.º de referencia del gen P07988
Alias de gen 18 kDa pulmonary-surfactant protein; 6 kDa protein; AI562151; P SFTB3; pot. surfactant-associated protein precursor; putative; preproprotein; PSP-B; Pulmonary surfactant protein 18; pulmonary surfactant protein B; pulmonary surfactant-associated protein B; pulmonary surfactant-associated protein B precursor; pulmonary surfactant-associated protein B; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SF-B; SFPB; SFTB3; SFTP3; Sftp-3; Sftpb; SMDP1; SP 18; SP-B; SPBS; surfactant associated protein B; surfactant protein B; surfactant protein-B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B
Símbolos de los genes SFTPB
Especie del huésped Mouse
Inmunógeno Recombinant Surfactant Protein B.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6439
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.