Descripción
- Obtain longer read lengths, more high-quality bases, and increased accuracy at the 5' end
- Get increased accuracy in regions with low signal-to-noise ratios or with anomalous signal artifacts such as spikes or dye blobs
- Determine the quality of your data using superior metrics from basecalling quality values
- Accelerate quality control using analysis reports with analysis statistics
- Filter out low-quality sequence ends automatically with sequence trimming
Longer Read Lengths with High-Quality Base Pairs
- Our significantly improved basecalling algorithm, the KB basecaller, now gives users up to 100 more high-quality bases than other basecalling algorithms
- Get longer read lengths with high-quality base pairs, mixed basecalling with quality value, and accurate basecalling of usually difficult-to-sequence short PCR fragments
Easily Review Sequencing Results with Quality Values
- This software enables users to customize and color code the range of the quality values to represent low-, medium-, and high-quality bases
- This way, when the basecaller identifies each base and assigns it a quality value, all you have to do is look at the color coding to easily review, discard, or accept it
- In addition, the software trims the ends of low-quality bases, grays them out on the user interface for easy identification, and calculates a sample score, which is the average quality value for all the bases in the untrimmed region
Reduce Data Screening Time
- Eliminate manual review of sequencing data batches
- With the software's Quality Control (QC) reports, get read length and sample score (average QV of bases in the clear range) for each sample file, enabling users to sort data by quality
- And to make reviewing data even easier, each QC report is hyperlinked back to its source data
Especificaciones
Especificaciones
| Software Type | Sequencing Data Analysis |
| Contenido y almacenamiento | Software/CD |
| Introducción de datos | Secuenciación CE (archivos .ab1) |
| Licencia | Inicial |
| Sistema operativo | Windows 10 Pro/IoT, Windows 7 Pro |
| Tipo de producto | Sequencing Analysis Software V7.0 Initial License |
| Cantidad | 1 licencia |
| Categoría de software | Admite archivos de todos los analizadores genéticos AB |