Learn More
Thermo Scientific™ Sensititre™ RAPMYCO2 Susceptibility Testing Plate
Código de producto. 18546944
Código de producto. 18546944 Proveedor Thermo Scientific™ N.º de proveedor YRAPMYCO2
Updated susceptibility testing for rapid growing mycobacterium which now includes Clofazimine, with high-quality, easy-to-use broth microdilution (BMD) plates, designed with reference to the latest CLSI guidelines (Research Use Only)
Perform susceptibility testing for rapid growing mycobacterium species (RGM) using broth microdilution, the recommended method for Antimicrobial Susceptibility Testing. The Thermo Scientific™ Sensititre™ RAPMYCO2 plate offers scientists an updated (from RAPMYCO) plate format that aligns with recently updated guidelines for susceptibility testing of nontuberculous mycobacteria.
Achieve reliable, consistent results for rapidly growing non-tuberculosis mycobacteria isolates with Sensititre RAPMYCO2 AST Plate.
  • Broth microdiltion (BMD) is the recommended AST method for mycobacteria
  • Test Mycobacterium spp. including rapid growing Mycobacterium fortuitum group, M. chelonaes group or M. abscessus group.
  • Designed with reference to the latest CLSI advice on drug options and accompanying breakpoints (where set)
  • BMD is a recommended AST method that maintains accurate results even where other methodologies fail
  • Extended shelf life, room-temperature storage eliminates inventory control concerns
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Easy set-up procedures and results in three days for rapid-growing organisms
  • Test 12 antimicrobials on a single AST plate for cost-effective results
Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.
The Sensititre System utilizes gold standard-level MIC accuracy1, providing superior quality and reproducibility for accurate results the first time. Broth microdilution is the reference method for AST, and in many cases2 the most reliable phenotypic method to guide therapeutic decisions.
TRUSTED_SUSTAINABILITY
Especificaciones
Agentes antimicrobianos Diverso
Descripción Sensititre Myco Susceptibility Plate – Rapid growing mycobacterium
Código de antibiótico Various
Tipo de producto AST Plate
Clase de organismo diana Rapidly-growing, Non-tuberculous Mycobacteria
Tipo de prueba Microbial Susceptibility
N.° por paquete 10
Tipo de empaquetado Box
Cantidad 10/Pk.
Especie Mycobacterium