Learn More
Cole-Parmer™ Sensidyne™ 800885-111-2301 GilAir™ 5 Sampling Pump Kit
Código de producto. 11919928 Tienda Cole-Parmer Productos
Encompass_Preferred
Cambiar vista
Click to view available options
Caudal:
De 0,75 a 3 L/min (rango de flujo bajo: de 1 a 500 mL/min)
Tamaño de la unidad:
cada uno
Código de producto. Caudal Tamaño de la unidad
11919928 De 0,75 a 3 L/min (rango de flujo bajo: de 1 a 500 mL/min) cada uno
1 options
Código de producto. 11919928 Proveedor Cole-Parmer™ N.º de proveedor 8627687
En stock
Ensure accuracy in high flow ranges with pumps featuring automated constant flow control. Cole-Parmer™ Sensidyne™ GilAir™ 5 Sampling Pump Kit includes pump, air boss (bag fill adapter), tubing, tool kit, rechargeable NiCd battery; 230VAC charger; and filter cassette holder.
  • EMI/FRI protection prevents interference from plant operations
  • See-through inlet filter housing protects pumps from dirt and debris while allowing easy inspection
  • GilAir 5 is a double-diaphragm pump with a larger capacity battery to handle higher flow rates
  • Clock features run and hold functions
Especificaciones
Incluye Pump, Air Boss (Bag Fill Adapter), Tubing, Tool Kit, Rechargeable NiCd Battery, 230 VAC Charger, Filter Cassette Holder
Certificaciones/conformidad UL (class I, groups A, B, C, D; class II, groups E, F, G; Class III, div 1); CENELEC
Tensión 230 VAC
Longitud global (imperial) 2 in.
Longitud global (métrico) 5.08 cm
Tipo de producto Air Sampling Pump
Caudal De 0,75 a 3 L/min (rango de flujo bajo: de 1 a 500 mL/min)
Altura total (imperial) 4 in.
Altura global (métrico) 10.16 cm
Anchura total (imperial) 3043/64 in.
Anchura global (métrico) 9.32 cm
Operating Temperature Range (English) 32° to 113°C
Operating Temperature Range (Metric) 0 to +40°C