missing translation for 'onlineSavingsMsg'
Learn More

HARPS Sempermed™ Green x-free NBR PF GR Gloves

Código de producto. 30274434
Change view
Click to view available options
Tamaño:
X-Small
Small
Medium
Large
X-Large
Tamaño de la unidad:
paquete de 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño unitSize
30274434 X-Large paquete de 200
30274433 X-Small paquete de 200
30274418 Small paquete de 200
30274432 Medium paquete de 200
30274428 Large paquete de 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30274434 Proveedor HARPS N.º de proveedor 3000016160

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Edge
Este artículo no se puede devolver. Vea la política de devoluciones

The sempermed™ green X-free is a accelerator-free with nitrile examination and personal protective glove.

  • SUSTAINABLE and SKIN-FRIENDLY: The sempermed™ green x-free is an accelerator-free, nitrile examination and personal protective glove.
  • Sempermed™ green X-free, a nitrile examination glove designed with both skin friendliness and environmental responsibility in mind.
  • Crafted with advanced nitrile latex it represents a significant step in our journey to offer the best solution to protect the skin of those who care for others, while following eco-friendly production practices.
  • The skin-friendly material minimises the risk of contact allergies as no rubber accelerators, sulphur, or silicones were used during production.
  • Ideal for users with Type I latex allergy and Type IV hypersensitivity, thanks to a skin-friendly formulation and minimal chemical residues.
  • Textured fingertips ensure secure handling and good tactile feedback, even with a lightweight design.
  • A silicone-free surface treatment allows for smooth and efficient glove changes in fast-paced settings.
  • With an AQL of 1.0, the gloves provide consistent barrier performance without compromising on sensitivity.
  • Protects users and patients from the transmission of infections.
  • Glove material is resistant to blood and other body fluids, and protects against bacteria, fungi and viruses including SARS-CoV-2, Hepatitis B and C and HIV.

Especificaciones

Chlorinated No
Accelerator Free Yes
Aplicaciones Healthcare
AQL 1
Color Green
Desechables Single-use
Acabado exterior Fingertip Textured
Finger Thickness (English) 3.1 mil
Finger Thickness (Metric) 0.08 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2 mil
Palm Thickness (Metric) 0.05 mm
En polvo Powder Free
Tipo de producto Nitrile Examination Glove
Tamaño X-Large
Esterilidad Non-sterile
Grosor (imperial) 2 mil
Grosor (métrico) 0.05 mm
FDA 510 (k) No
Estilo de puño de los guantes Beaded/Rolled
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.