missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEL1L3 Polyclonal Antibody

Description
Immunogen sequence: ERCAEVQEIVS VYASAAKHGG ERQEACHLHN SYLDLQRRYG RPSMCRAFPW EKELKDKHPS LFQALLEMDL LTVPRNQNES VSEIGGKIFE KAVKRLSSID GL Highest antigen sequence identity to the following orthologs - mouse 70%, rat 67%.
SEL1L3 is a protein coding gene.
Specifications
Specifications
| Antígeno | SEL1L3 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | SEL1L3 |
| N.º de referencia del gen | Q68CR1 |
| Alias de gen | KIAA0746; Protein sel-1 homolog 3; sel-1 suppressor of lin-12-like 3; SEL1L family member 3; SEL1L3; Sel-1L3; Suppressor of lin-12-like protein 3 |
| Símbolos de los genes | SEL1L3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?