missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Securin Monoclonal Antibody (K49008_2B9), Invitrogen™

Código de producto. 30209759
Change view
Click to view available options
Cantidad:
50 μL
200 μL
Tamaño de la unidad:
200 microgramos
50 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30209759 200 μL 200 microgramos
30202429 50 μL 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30209759 Proveedor Invitrogen™ N.º de proveedor MA556155

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MATLIYVDKE NGEPGTRVVA KDGLKLGSGP SIKALDGRSQ VSTPRFGKTF DAPPALPKAT RKALGTVNRA TEKSVKTKGP LKQKQPSFSA KKMTEKTVKA KSSVPASDDA YPEIEKFFPF NPLDFESFDL PEEHQIAHLP LSGVPLMILD EERELEKLFQ LGPPSPVKMP SPPWESNLLQ SPSSILSTLD VELPPVCCDI DI

The encoded protein is a homolog of yeast securin proteins, which prevent separins from promoting sister chromatid separation. It is an anaphase-promoting complex (APC) substrate that associates with a separin until activation of the APC. The gene product has transforming activity in vitro and tumorigenic activity in vivo, and the gene is highly expressed in various tumors. The gene product contains 2 PXXP motifs, which are required for its transforming and tumorigenic activities, as well as for its stimulation of basic fibroblast growth factor expression. It also contains a destruction box (D box) that is required for its degradation by the APC. The acidic C-terminal region of the encoded protein can act as a transactivation domain. The gene product is mainly a cytosolic protein, although it partially localizes in the nucleus.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Securin
Aplicaciones Western Blot
Clasificación Monoclonal
Clon K49008_2B9
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
génica Pttg1
N.º de referencia del gen O95997
Alias de gen AW555095; C87862; EAP1; Esp1-associated protein; ESP1-associated protein 1; HPTTG; MGC126883; MGC138276; Pituitary tumor transforming; pituitary tumor-transforming 1; pituitary tumor-transforming gene 1; pituitary tumor-transforming gene 1 protein; PTTG; PTTG1; Pttg3; Securin; tumor-transforming protein 1; TUTR1
Símbolos de los genes Pttg1
Especie del huésped Mouse
Inmunógeno Recombinant human PTTG1 (immunogen range is 1-202).
Método de purificación Protein A
Cantidad 200 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9232
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.