missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Securin Monoclonal Antibody (K49008_2B9), Invitrogen™
Descripción
Sequence of this protein is as follows: MATLIYVDKE NGEPGTRVVA KDGLKLGSGP SIKALDGRSQ VSTPRFGKTF DAPPALPKAT RKALGTVNRA TEKSVKTKGP LKQKQPSFSA KKMTEKTVKA KSSVPASDDA YPEIEKFFPF NPLDFESFDL PEEHQIAHLP LSGVPLMILD EERELEKLFQ LGPPSPVKMP SPPWESNLLQ SPSSILSTLD VELPPVCCDI DI
The encoded protein is a homolog of yeast securin proteins, which prevent separins from promoting sister chromatid separation. It is an anaphase-promoting complex (APC) substrate that associates with a separin until activation of the APC. The gene product has transforming activity in vitro and tumorigenic activity in vivo, and the gene is highly expressed in various tumors. The gene product contains 2 PXXP motifs, which are required for its transforming and tumorigenic activities, as well as for its stimulation of basic fibroblast growth factor expression. It also contains a destruction box (D box) that is required for its degradation by the APC. The acidic C-terminal region of the encoded protein can act as a transactivation domain. The gene product is mainly a cytosolic protein, although it partially localizes in the nucleus.
Especificaciones
Especificaciones
| Antígeno | Securin |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | K49008_2B9 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| génica | Pttg1 |
| N.º de referencia del gen | O95997 |
| Alias de gen | AW555095; C87862; EAP1; Esp1-associated protein; ESP1-associated protein 1; HPTTG; MGC126883; MGC138276; Pituitary tumor transforming; pituitary tumor-transforming 1; pituitary tumor-transforming gene 1; pituitary tumor-transforming gene 1 protein; PTTG; PTTG1; Pttg3; Securin; tumor-transforming protein 1; TUTR1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?