Learn More
Tocris Bioscience™ SCOTfluor 510, fluoro
Descripción
SCOTfluor 510, fluoro is a precursor that, upon reaction with amine groups, can be used for the synthesis of fluorescent probes. Can be used to generate functionalized lactic acid, glucose and ceramide fluorescent probes for real-time, in situ imaging of small molecule metabolites in vivo and in live cells. Fluorescent adduct excitation maximum = 510 nm; emission maximum = 606 nm.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Fluoro-7-nitro-2,1,3-benzoselenadiazole |
| CAS | 2351940-09-3 |
| Cantidad | 50 mg |
| Objetivo | Other Fluorescent Probes |
| Fórmula molecular | C6H2FN3O2Se |
| Pureza | 0.9 |