Learn More
Schisandrin, MedChemExpress
Código de producto. 16439068
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16439068 10 mg 10 miligramos
16404877 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16439068 Proveedor Medchem Express N.º de proveedor HYN0691
MedChemExpress Schisandrin (Schizandrin), a dibenzocyclooctadiene lignan, is isolated from the fruit of Schisandra chinensis Baill. Schisandrin exhibits antioxidant, hepatoprotective, anti-cancer and anti-inflammatory activities. Schisandrin also can reverses memory impairment in rats.
Schisandrin (Schizandrin), a dibenzocyclooctadiene lignan, is isolated from the fruit of Schisandra chinensis Baill. Schisandrin exhibits antioxidant, hepatoprotective, anti-cancer and anti-inflammatory activities. Schisandrin also can reverses memory impairment in rats.
Especificaciones
Nombre del producto químico o material Schisandrin
CAS 7432-28-2
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C24H32O7
Cantidad 10 mg
Sinónimo SchizandrinSchizandrol Schizandrol-A
Información de solubilidad DMSO : 1 mg/mL (2.31 mM; Need ultrasonic)
SMILES O[C@]1(C)[C@@H](C)CC2=CC(OC)=C(OC)C(OC)=C2C3=C(OC)C(OC)=C(OC)C=C3C1
Peso molecular (g/mol) 432.51
Formula Weight (peso de la fórmula) 432.51
Porcentaje de pureza 99.51%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation